Would you like to log in again?
Anti-Cytokeratin 8 Mouse Monoclonal Antibody [clone: 35bH11]
Supplier: Diagnostic BioSystems
PDM117-RUO PDM117
76634-364EA 231.69 USD InStock
76634-364 CA76634-364
Anti-Cytokeratin 8 Mouse Monoclonal Antibody [clone: 35bH11]
Antibodies
This antibody recognizes keratin polypeptide of 54 kDa. It stains strongly with non-squamous epithelium but does not stain squamous epithelium. It reacts with the sweat gland and ducts in the skin. Adenocarcinomas of the ovary, gastrointestinal tract, ductal carcinoma of breast, pancreas and bile ducts, stain positive with this antibody.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty