Would you like to log in again?
Anti-EBV cocktail Mouse Monoclonal Antibody [clone: CS1, CS2, CS3, CS4]
Supplier: Diagnostic BioSystems
PDM128-RUO PDM128
76634-380EA 279.47 USD InStock
CA76634-380 76634-380
Anti-EBV cocktail Mouse Monoclonal Antibody [clone: CS1, CS2, CS3, CS4]
Antibodies
This antibody is specific to 60 kDa latent membrane protein (LMP-1) encoded by the BNLF1 gene of the EBV. Each clone reacts with different epitopes on the hydrophilic C-terminus of the cytoplasmic domain of LMP-1. This antibody stains strongly with EBV positive lymphoblastoid cell lines and EBV infected B cell immunoblasts in infectious mononucleosis.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty