Would you like to log in again?
Anti-Mesothelioma Mouse Monoclonal Antibody [clone: HBME-1]
Supplier: Diagnostic BioSystems
PDM134 PDM134-RUO
76634-386EA 536.24 USD InStock
76634-386 CA76634-386
Anti-Mesothelioma Mouse Monoclonal Antibody [clone: HBME-1]
Antibodies
This antibody reacts with an unknown antigen on microvilli of mesothelioma cells. It stains normal mesothelial cells as well as epithelial mesotheliomas in a thick membrane pattern due to abundant lung microvilli on the surface of these cells.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty