Would you like to log in again?
Anti-Cytokeratin 5 and 14 Mouse Monoclonal Antibody [clone: XM26 + LL002]
Supplier: Diagnostic BioSystems
PDM140-RUO PDM140
76634-392EA 332.01 USD InStock
CA76634-392 76634-392
Anti-Cytokeratin 5 and 14 Mouse Monoclonal Antibody [clone: XM26 + LL002]
Antibodies
This antibody cocktail reacts with cytokeratin 5 intermediate filament protein as well as the 50 kDa cytokeratin protein designated cytokeratin 14. Basal cells of human epidermis express acidic cytokeratin 14 and basic cytokeratin 5. Cytokeratin 5 is a 58 kDa protein, which is closely related to cytokeratin 6. Cytokeratin 5 and 6 are expressed in basal cell epithelioma, basal cells of prostate, urothelium, vagina, squamous cell carcinomas of skin, tongue, epiglottis and rectal-anal region. Cytokeratin 5 can be useful in the distinction of mesotheliomas from most adenocarcinomas.Cytokeratin 14 belongs to the type A (acidic) subfamily of low MW cytokeratins and exists in combination with cytokeratin 15. This antibody can be useful to distinguish stratified epithelial cells from simple epithelial cells.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty