Would you like to log in again?
Anti-CD25 Mouse Monoclonal Antibody [clone: IL2R.1]
Supplier: Diagnostic BioSystems
PDM144-RUO PDM144
76634-398EA 290.21 USD InStock
76634-398 CA76634-398
Anti-CD25 Mouse Monoclonal Antibody [clone: IL2R.1]
Antibodies
This antibody is specific to a 55 kDa protein known as interleukin 2-receptor α (IL-2R α). IL-2R α is over expressed on the surface of activated T cells, HTLV-1 or HTLV-2 positive T cell lines and moderately expressed on activated B cells, NK-cells and macrophages.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty