Would you like to log in again?
Anti-Myogenin Mouse Monoclonal Antibody [clone: F5D]
Supplier: Diagnostic BioSystems
PDM158-RUO PDM158
76634-420EA 339.18 USD InStock
76634-420 CA76634-420
Anti-Myogenin Mouse Monoclonal Antibody [clone: F5D]
Antibodies
This antibody is specific to 34 kDa protein known as myogenin. Myogenin is a member of family of myogenic regulatory genes which includes MyoD, MyF5 and MRF4. These genes encode a set of transcription factors which are essential for muscle development. Expression of myogenin is restricted to cells of skeletal muscle origin.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty