Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Anti-Wilms Tumor 1 protein (WT1) Mouse Monoclonal Antibody [clone: 6F-H2]
Supplier: Diagnostic BioSystems
PDM177 PDM177-RUO
76634-452EA 394.12 USD InStock
76634-452 CA76634-452
Anti-Wilms Tumor 1 protein (WT1) Mouse Monoclonal Antibody [clone: 6F-H2]
Antibodies
Wilms tumor-1 (WT-1), is a gene involved in the induction of Wilms tumor, a pediatric renal malignancy. Wilms tumor is associated with mutations of WT1, a zinc-finger transcription factor that is essential for the development of the metanephric kidney and the urogenital system. The WT1 gene is normally expressed in fetal kidney and mesothelium, and its expression has been suggested as a marker for Wilms tumor and mesothelioma.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty