Would you like to log in again?
Anti-Cytokeratin 17 Mouse Monoclonal Antibody [clone: E3]
Supplier: Diagnostic BioSystems
PDM206 PDM206-RUO
76634-474EA 279.47 USD InStock
76634-474 CA76634-474
Anti-Cytokeratin 17 Mouse Monoclonal Antibody [clone: E3]
Antibodies
This antibody is specific to 46 kDa cytokeratin protein designated cytokeratin 17. It reacts with basal cells in complex epithelia and reacts strongly with squamous cell carcinomas.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty