Would you like to log in again?
Anti-Bcl-10 Mouse Monoclonal Antibody [clone: 151]
Supplier: Diagnostic BioSystems
PDM442 PDM442-RUO
76634-512EA 287.83 USD InStock
76634-512 CA76634-512
Anti-Bcl-10 Mouse Monoclonal Antibody [clone: 151]
Antibodies
Monoclonal anti-bcl-10 reacts specifically with human bcl-10. The epitope recognized by the antibody resides within amino acids 122-168 of human bcl-10 molecule. Bcl-10, also designated as CIPER, mE10, cE10, CARMEN, and CLAP, is an N-terminal CARD (caspase recruitment domain) containing protein. It is a cellular homologue of the equine herpesvirus-2 protein E-10 (vCLAP). Bcl-10 was implicated in the regulation of apoptosis by interacting with caspase 9, enhancing procaspase 9 processing, and triggering its activation when overexpressed in the cell. Bcl-10 cellular overexpression induces JNK, p38, and NF-kB activation. Deregulation of bcl-10 expression was also demonstrated to be involved in cellular oncogenesis.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty