Would you like to log in again?
Anti-Galectin-3 Mouse Monoclonal Antibody [clone: H-5]
Supplier: Diagnostic BioSystems
PDM541 PDM541-RUO
76634-516EA 385.76 USD InStock
76634-516 CA76634-516
Anti-Galectin-3 Mouse Monoclonal Antibody [clone: H-5]
Antibodies
Galectin-3 is a member of the beta-galactoside-binding protein family that plays an important role in cell adhesion, cell matrix interactions, macrophage activation, angiogenesis, metastasis, and apoptosis.
Galectin-3 shows strong diffuse cytoplasmic staining in most cases of papillary thyroid carcinoma (PTC), most poorly differentiated thyroid cancers demonstrate galectin-3 protein expression while it is not expressed in anaplastic thyroid cancers. (1) Upregulated expression of galectin-3 is involved in cancer progression and metastasis, and it is suggested that Galectin-3 may be a valuable tool to support diagnosis of parathyroid carcinoma. (2)
Galectin-3 has also been reported to play important roles in the progression of prostate cancer, and their heterogeneous expressions might be associated with different cancer subtypes. (3)
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty