Would you like to log in again?
Anti-Herpes simplex virus II (HSV II) Mouse Monoclonal Antibody [clone: DBM15.69]
Supplier: Diagnostic BioSystems
PDM542 PDM542-RUO
76634-518EA 213.78 USD InStock
CA76634-518 76634-518
Anti-Herpes simplex virus II (HSV II) Mouse Monoclonal Antibody [clone: DBM15.69]
Antibodies
This antibody reacts with herpes simplex virus type 2. It is specific for the viral glycoprotein D (gD) protein.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty