Would you like to log in again?
Anti-Podoplanin Mouse Monoclonal Antibody [clone: D2-40]
Supplier: Diagnostic BioSystems
PDM558 PDM558-RUO
76634-530EA 526.68 USD InStock
76634-530 CA76634-530
Anti-Podoplanin Mouse Monoclonal Antibody [clone: D2-40]
Antibodies
Podoplanin is a 38 kDa membrane mucoprotein originally detected on the surface of rat podocytes .The antibody against podoplanin (D2-40) reacts to a 40 kDA sialogylcoprotein found on lymphatic endothelium, fetal testis, and on the surface of testicular germ cell tumors. Podoplanin can be used to assist pathologists in identifying lymphatic endothelium to determine lymphatic invasion and also can be used to help differentiate mesothelioma from adenocarcinoma.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty