Would you like to log in again?
Anti-Cytokeratin 7 Mouse Monoclonal Antibody [clone: LP1K]
Supplier: Diagnostic BioSystems
PDM563-RUO PDM563
76634-538EA 311.72 USD InStock
76634-538 CA76634-538
Anti-Cytokeratin 7 Mouse Monoclonal Antibody [clone: LP1K]
Antibodies
This antibody recognizes keratin polypeptide of 54 kDa. It stains a number of epithelial cells including ductal and glandular epithelia. Stratified squamous epithelia are positive but transitional epithelium of the urinary tract does not show any staining. This antibody labels adenocarcinomas and transitional carcinomas but does not react with squamous cell carcinomas.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty