Would you like to log in again?
Anti-CA 19-9 Mouse Monoclonal Antibody [clone: C241:5:1:4]
Supplier: Diagnostic BioSystems
PDM567 PDM567-RUO
76634-546EA 271.11 USD InStock
CA76634-546 76634-546
Anti-CA 19-9 Mouse Monoclonal Antibody [clone: C241:5:1:4]
Antibodies
This antibody reacts specifically with sialyl Lewisa and recognizes an epitope being designated CA 19-9. It does not cross-reacts with Lewisa and Lewisb. CA 19-9 has been shown to be useful marker in the diagnosis and management of gastrointestinal cancers.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty