Would you like to log in again?
Anti-PTEN Mouse Monoclonal Antibody [clone: 6H2.1]
Supplier: Diagnostic BioSystems
PDM574 PDM574-RUO
76634-558EA 297.38 USD InStock
CA76634-558 76634-558
Anti-PTEN Mouse Monoclonal Antibody [clone: 6H2.1]
Antibodies
PTEN gene is a tumor suppressor gene that maps to chromosome 10q23. PTEN, a novel tumor suppressor, functions as a regulator of both cell cycle progression and apoptosis ). Potentially, mutation and deletion of PTEN gene may result in a new signal transduction pathway related to human malignant tumors. Studies have demonstrated a reduction of PTEN expression in advanced breast cancers.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty