Would you like to log in again?
Anti-Neuron specific enolase Mouse Polyclonal Antibody
Supplier: Diagnostic BioSystems
PDR006 PDR006-RUO
76634-608EA 205.42 USD InStock
76634-608 CA76634-608
Anti-Neuron specific enolase Mouse Polyclonal Antibody
Antibodies
This antibody is specific to γ-γ enolase isozyme of 46 kDa. This antibody stains neurons, neuroendocrine cells, gangliomas, oat cell carcinomas, astrocytomas, paragangliomas, glioblastomas, pheochromocytomas, melanomas and schwannomas.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty