Would you like to log in again?
Anti-Factor VIII R antigen Rabbit Polyclonal Antibody
Supplier: Diagnostic BioSystems
PDR014 PDR014-RUO
76634-614EA 280.66 USD InStock
76634-614 CA76634-614
Anti-Factor VIII R antigen Rabbit Polyclonal Antibody
Antibodies
This antibody reacts with human factor VIII related antigen. It stains endothelial cells and megakaryocytes in various human tissues. This antibody does not cross-react with any other cell types.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty