Would you like to log in again?
Anti-Kappa light chain Rabbit Polyclonal Antibody
Supplier: Diagnostic BioSystems
PDR015-RUO PDR015
76634-616EA 231.69 USD InStock
CA76634-616 76634-616
Anti-Kappa light chain Rabbit Polyclonal Antibody
Antibodies
This antibody reacts with free as well as bound kappa light chains. Contaminating antibodies have been removed by solid phase adsorption.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty