Would you like to log in again?
Anti-Estrogen Receptor (ER) Rabbit Monoclonal Antibody [clone: SP1]
Supplier: Diagnostic BioSystems
RMAB001-RUO RMAB001-05-RUO RMAB001-01 RMAB001-01-RUO RMAB001-05 RMAB001
76634-694EA 2257.2 USD InStock
CA76634-692 76634-694 CA76634-694 76634-690 CA76634-690 76634-692
Anti-Estrogen Receptor (ER) Rabbit Monoclonal Antibody [clone: SP1]
Antibodies
This antibody recognizes a protein of 67 kDa, which is identified as estrogen receptor (ER). ER content of breast cancer tissue is an important parameter in the prediction of prognosis and response to endocrine therapy. This antibody strongly stains the nucleus of epithelial cells in breast carcinomas. The ER is an important regulator of growth and differentiation in the mammary gland. Presence of ER in breast tumors indicates an increased likelihood of response to anti-estrogen (e.g. tamoxifen) therapy.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty