Would you like to log in again?
Anti-CD23 B Cell Rabbit Monoclonal Antibody [clone: SP23]
Supplier: Diagnostic BioSystems
RMAB013-01-RUO RMAB013-05-RUO RMAB013-05 RMAB013 RMAB013-01 RMAB013-RUO
76634-746EA 702.24 USD InStock
CA76634-748 CA76634-744 76634-746 CA76634-746 76634-748 76634-744
Anti-CD23 B Cell Rabbit Monoclonal Antibody [clone: SP23]
Antibodies
This antibody recognizes CD23. CD23 is a 45 kDa glycoprotein, which is present on a subpopulation of freshly isolated peripheral blood and tonsil B cells and strongly expressed on EBV-transformed B lymphoblasts. The CD23 molecule is identical to the low affinity IgE receptor found on B cells. Expression of CD23 has been detected in neoplastic cells from cases of B cell chronic lymphocyctic leukemia and some cases of centroblastic/centrocytic lymphoma.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty