Would you like to log in again?
Anti-Chromogranin A Rabbit Monoclonal Antibody [clone: SP12]
Supplier: Diagnostic BioSystems
RMAB015-05 RMAB015-01 RMAB015 RMAB015-05-RUO RMAB015-RUO RMAB015-01-RUO
76634-750EA 154.07 USD InStock
76634-750 76634-752 CA76634-750 76634-754 CA76634-752 CA76634-754
Anti-Chromogranin A Rabbit Monoclonal Antibody [clone: SP12]
Antibodies
Chromogranin A is a 439 amino acid protein present in secretory granules of a wide variety of endocrine cells and neurons. A positive staining is seen in the secretory granules of parathyroid, adrenal medulla, anterior pituitary gland and Langerhans islets of pancreas. This antibody stains a variety of neuroendocrine tumors.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty