Would you like to log in again?
Anti-PLAP Rabbit Monoclonal Antibody [clone: SP15]
Supplier: Diagnostic BioSystems
RMAB017-05-RUO RMAB017-01-RUO RMAB017-RUO RMAB017-05 RMAB017 RMAB017-01
76634-760EA 974.54 USD InStock
76634-760 CA76634-760 CA76634-756 76634-758 CA76634-758 76634-756
Anti-PLAP Rabbit Monoclonal Antibody [clone: SP15]
Antibodies
This antibody reacts with a membrane-bound isoenzyme (Regan and Nagao type) of placental alkaline phosphatase (PLAP) occurring in the placenta during the 3rd trimester of gestation. This antibody is highly specific to PLAP and shows no cross-reaction with other isoenzymes of alkaline phosphatases. It is useful in the identification of testicular germ cell tumors. Unlike germ cell tumors, PLAP-positive somatic cell tumors uniformly express epithelial membrane antigen (EMA).
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty