Would you like to log in again?
Anti-Synaptophysin Rabbit Monoclonal Antibody [clone: SP11]
Supplier: Diagnostic BioSystems
RMAB018-01 RMAB018-01-RUO RMAB018-05-RUO RMAB018-05 RMAB018-RUO RMAB018
76634-762EA 154.07 USD InStock
76634-762 CA76634-766 CA76634-762 76634-764 CA76634-764 76634-766
Anti-Synaptophysin Rabbit Monoclonal Antibody [clone: SP11]
Antibodies
This antibody is specific to a 38 kDa protein. Synaptophysin is an glycoprotein occurring in presynaptic vesicles of neurons in the brain, spinal cord, retina, vesicles of adrenal medulla and in neuromuscular junctions. This antibody reacts with neuroendocrine neoplasms of neural as well as epithelial types.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty