Would you like to log in again?
Anti-Mammoglobin A Rabbit Monoclonal Antibody [clone: EPR 9092]
Supplier: Diagnostic BioSystems
RMAB036 RMAB036-05 RMAB036-05-RUO RMAB036-RUO
76634-770EA 644.92 USD InStock
CA76634-770 76634-770 76634-768 CA76634-768
Anti-Mammoglobin A Rabbit Monoclonal Antibody [clone: EPR 9092]
Antibodies
The mammaglobin gene was first identified using a differential screening approach directed at the isolation of novel, human breast cancer-associated genes. Mammaglobin-A is highly overexpressed in breast cancer cell lines and primary breast tumors. This pattern of expression is restricted to mammary epithelium and metastatic breast tumors. Thus, mammaglobin-A-specific T cell immune responses may provide an important approach for the design of breast cancer-specific immunotherapy.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty