Would you like to log in again?
Anti-CDX2 Rabbit Monoclonal Antibody [clone: EP25]
Supplier: Diagnostic BioSystems
RMAB059-05-RUO RMAB059-01-RUO RMAB059-05-RUO RMAB059 RMAB059-RUO RMAB059-01
76634-784EA 198.25 USD InStock
76634-784 CA76634-788 76634-786 CA76634-784 CA76634-786 76634-788
Anti-CDX2 Rabbit Monoclonal Antibody [clone: EP25]
Antibodies
The caudal-related homeodomain protein 2, CDX-2, is a transcription factor which is expressed in the intestine and is thought to play an important role in the proliferation and differentiation of intestinal epithelial cells. The CDX-2 protein is expressed in primary and metastatic colorectal carcinomas, intestinal metaplasia of the stomach and intestinal type gastric cancer. In human colorectal cancer, the expression of both CDX2 and carbonic anhydrase 1, a gene regulated by CDX2, is reduced or absent. CDX-2 is one of the important regulators in defining pathways for coordinate control of drug metabolism in the gastrointestinal tract.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty