Would you like to log in again?
Anti-P504S Rabbit Monoclonal Antibody [clone: 13H4]
Supplier: Diagnostic BioSystems
RMAB078-01 RMAB078-RUO RMAB078-RUO RMAB078-05-RUO RMAB078-05-RUO RMAB078-01-RUO
76634-796EA 249.61 USD InStock
76634-796 CA76634-800 76634-798 CA76634-796 76634-800 CA76634-798
Anti-P504S Rabbit Monoclonal Antibody [clone: 13H4]
Antibodies
P504S has been recently described as a prostate cancer-specific gene that encodes a protein involved in the β-oxidation of branched chain fatty acids. Expression of p504S protein is found in prostatic adenocarcinoma but not in benign prostatic tissue. It stains premalignant lesions of prostate: high-grade prostatic intraepithelial neoplasia (PIN) and atypical adenomatous hyperplasia. P504S can be used as a positive marker for PIN.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty