Would you like to log in again?
Anti-CD4 Rabbit Monoclonal Antibody [clone: EP204]
Supplier: Diagnostic BioSystems
RMAB083-05 RMAB083-01-RUO RMAB083-01-RUO RMAB083 RMAB083-RUO RMAB083-05-RUO
76634-806EA 1006.79 USD InStock
76634-806 CA76634-804 CA76634-806 76634-802 76634-804 CA76634-802
Anti-CD4 Rabbit Monoclonal Antibody [clone: EP204]
Antibodies
CD4 is glycoprotein found on the surface of immune cells such as T helper cells, monocytes, macrophages, and dendritic cells. It is a co-receptor that assists the T-cell receptor (TCR) with antigen-presenting cell and also interacts directly with MHC class II molecules of the surface of antigen presenting cells. Most mature T-cell lymphomas are CD4 positive with the exception of aggressive NK-cell leukemia and extranodal NL/T-cell lymphoma. CD4 plays an important role in the classification of lymphocytes in inflammatory legions and malignant lymphomas.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty