Would you like to log in again?
Anti-P63 Rabbit Monoclonal Antibody [clone: DBR16.1]
Supplier: Diagnostic BioSystems
RMAB086-05-RUO RMAB086-RUO RMAB086-01-RUO RMAB086-05 RMAB086-01 RMAB086
76634-808EA 130.18 USD InStock
CA76634-808 76634-808 CA76634-810 76634-812 CA76634-812 76634-810
Anti-P63 Rabbit Monoclonal Antibody [clone: DBR16.1]
Antibodies
p63 is a homolog of the tumor suppressor p53, it is identified in basal cells in the epithelial layers of a variety of tissues, including epidermis, cervix, urothelium, breast and prostate. p63, vital for the development of the prostate and selectively expressed by normal prostate basal cells, has been shown to be a promising complementary basal cell specific marker to High Molecular Weight-Cytokeratin (HMW-CK), for the differential diagnosis of benign prostatic lesions and prostatic carcinoma. p63 has also been shown to be a sensitive marker for lung squamous cell carcinomas (SqCC), with a sensitivity of ~90% . Specificity for lung SqCC, vs. lung adenocarcinoma (LADC), is approximately 80. In breast tissue, p63 has been identified in myoepithelial cells of normal ducts.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty