Would you like to log in again?
Anti-PRAME Rabbit Monoclonal Antibody [clone: EPR20330]
Supplier: Diagnostic BioSystems
RMAB109 RMAB109-01-RUO RMAB109-05 RMAB109-RUO RMAB109-01 RMAB109-05-RUO
76634-828EA 1138.16 USD InStock
CA76634-826 76634-828 CA76634-828 76634-826 76634-830 CA76634-830
Anti-PRAME Rabbit Monoclonal Antibody [clone: EPR20330]
Antibodies
PRAME (Preferentially expressed Antigen in Melanoma) is a tumorassociated antigen and is a member of the family of cancer testis antigens (CTA). PRAME is expressed in malignant cells, including leukaemias, Hodgkin's lymphoma, breast cancer, and primary and metastatic melanomas. PRAME has low or no expression in normal tissues except for in testis, ovary, placenta, adrenals and endometrium.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty