Would you like to log in again?
Anti-Progesterone receptor (PR) Rabbit Monoclonal Antibody [clone: SP2]
Supplier: Diagnostic BioSystems
RMPD002-RUO RMPD002
76634-840EA 789.43 USD InStock
76634-840 CA76634-840
Anti-Progesterone receptor (PR) Rabbit Monoclonal Antibody [clone: SP2]
Antibodies
The progesterone receptor (PgR) is an estrogen-regulated protein. It has been proposed that expression of PgR indicates a responsive estrogen receptor (ER) pathway. A number of studies have shown that PgR determination provides supplementary information to ER, both in predicting response to endocrine therapy and estimating survival. PgR has proved superior to ER as a prognostic indicator in some studies.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty