Would you like to log in again?
Anti-Cyclin D1 Rabbit Monoclonal Antibody [clone: SP4]
Supplier: Diagnostic BioSystems
RMPD003-RUO RMPD003
76634-842EA 609.09 USD InStock
76634-842 CA76634-842
Anti-Cyclin D1 Rabbit Monoclonal Antibody [clone: SP4]
Antibodies
Cyclin D1 or PRAD-1 or bcl-1 is one of the key cell cycle regulators, and functions in association with cdk4 and/or cdk6 by phosphorylating the Rb protein. It is a putative proto-oncogene overexpressed in a wide variety of human neoplasms including mantle cell lymphomas (MCL).
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty