Would you like to log in again?
Anti-Ki-67 antigen Rabbit Monoclonal Antibody [clone: SP6]
Supplier: Diagnostic BioSystems
RMPD004-RUO RMPD004
76634-844EA 474.13 USD InStock
CA76634-844 76634-844
Anti-Ki-67 antigen Rabbit Monoclonal Antibody [clone: SP6]
Antibodies
Ki-67 is a nuclear protein, which is expressed in proliferating cells. Ki-67 is preferentially expressed during late G1-, S-, M-, and G2-phases of the cell cycle, while cells in the G0 (quiescent) phase are negative for this protein.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty