Would you like to log in again?
Anti-HER2/neu (c-erbB-2) Rabbit Monoclonal Antibody [clone: SP3]
Supplier: Diagnostic BioSystems
RMPD008 RMPD008-RUO
76634-848EA 732.1 USD InStock
CA76634-848 76634-848
Anti-HER2/neu (c-erbB-2) Rabbit Monoclonal Antibody [clone: SP3]
Antibodies
This antibody recognizes a c-erbB-2 protein, which is a receptor tyrosine kinase of the c-erbB family. It is closely related in structure to the epidermal growth factor receptor. c-erbB-2 oncoprotein is detectable in a subset of breast and other adenocarcinomas, as well as transitional cell carcinomas. In the case of breast cancer, expression determined by immunohistochemistry has been shown to be associated with poor prognosis.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty