Would you like to log in again?
Anti-CD5 Rabbit Monoclonal Antibody [clone: SP19]
Supplier: Diagnostic BioSystems
RMPD011-RUO RMPD011
76634-852EA 610.28 USD InStock
76634-852 CA76634-852
Anti-CD5 Rabbit Monoclonal Antibody [clone: SP19]
Antibodies
This antibody recognizes CD5, which is a transmembrane protein, is found on 95% of thymocytes and 72% of peripheral blood lymphocytes. In lymph nodes, the main reactivity is observed in T cell areas. CD5 is expressed by many T cell leukemia, lymphomas, and activated T cells. Occasionally, CD5 antigen is also expressed on a subset of B cells. Mantle cell lymphomas (same as diffuse centrocytic lymphomas) are CD5(+) while the follicle center cell lymphoma is CD5(-).
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty