Would you like to log in again?
Anti-CD23 Rabbit Monoclonal Antibody [clone: SP23]
Supplier: Diagnostic BioSystems
RMPD013-RUO RMPD013
76634-856EA 610.28 USD InStock
76634-856 CA76634-856
Anti-CD23 Rabbit Monoclonal Antibody [clone: SP23]
Antibodies
This antibody is specific to a membrane glycoprotein of 45 kDa, designated as CD23. CD23 is found on a subpopulation of peripheral blood cells, B lymphocytes and on EBV transformed B lymphoblastoid cell lines. Expression of CD23 has been detected in neoplastic cells from cases of B cell chronic lymphocytic leukemias.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty