Would you like to log in again?
Anti-SOX-10 (melanoma marker) Rabbit Monoclonal Antibody [clone: EP268]
Supplier: Diagnostic BioSystems
RMPD077 RMPD077-RUO
76634-874EA 252 USD InStock
76634-874 CA76634-874
Anti-SOX-10 (melanoma marker) Rabbit Monoclonal Antibody [clone: EP268]
Antibodies
SOX-10, a member of the sex-determining region Y-related HMG-box family, is a transcription factor that is expressed in pre-migratory and migrating neural crest cells. Following migration it remains expressed in cells destined for glial, schwannian, and melanocytic differentiation. SOX10 is widely expressed in normal human tissues including melanocytes and breast tissue. SOX10 is also an important marker in malignant tumors such as melanoma, and breast carcinomas of basal-like and triple negative type, gliomas, and benign tumors such as schwannomas. More importantly, SOX10 has been shown to be expressed in 97 to 100% of desmoplastic and spindle cell melanomas and has also been shown to be expressed in 100% of nevi. Spindle cell and desmoplastic melanomas are rare variants of invasive cutaneous melanoma, with an annual incidence rate of approximately 2 per 100,000. The majority of oligodendrogliomas and a large percentage of astrocytomas and poorly differentiated glioblastomas have also been shown to express SOX10.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty