Would you like to log in again?
Anti-ROS1 Rabbit Monoclonal Antibody [clone: EPMGHR2]
Supplier: Diagnostic BioSystems
RMPD108 RMPD108-RUO
76634-886EA 1053.36 USD InStock
76634-886 CA76634-886
Anti-ROS1 Rabbit Monoclonal Antibody [clone: EPMGHR2]
Antibodies
ROS1 rearrangements occur infrequently in lung ACA, however given the frequency of lung cancer in the population, ROS1-rearranged tumors represent a significant number of cancer patients. ROS1 gene rearrangements are reported in 1 to 2% of lung adenocarcinomas (ACA) and are associated with response to the multitargeted tyrosine kinase inhibitor, crizotinib. ROS IHC can be readily incorporated into the diagnostic surgical pathology workup of lung adenocarcinoma, with results confirmed by FISH as needed.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty