Would you like to log in again?
Anti-BOB.1 Rabbit Monoclonal Antibody [clone: SP92]
Supplier: Diagnostic BioSystems
RMPD110-RUO RMPD110
76634-890EA 292.6 USD InStock
CA76634-890 76634-890
Anti-BOB.1 Rabbit Monoclonal Antibody [clone: SP92]
Antibodies
Transcriptional coactivator that specifically associates with either OCT1 or OCT2. It boosts the OCT1 mediated promoter activity and to a lesser extent, that of OCT2. It has no intrinsic DNAbinding activity. It recognizes the POU domains of OCT1 and OCT2. It is essential for the response of B-cells to antigens and required for the formation of germinal centers.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty