Would you like to log in again?
Anti-HER2/neu (c-erb-2) Rabbit Polyclonal Antibody
Supplier: Diagnostic BioSystems
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
RP006-01 RP006-05-RUO RP006-05-RUO RP006-01-RUO RP006-RUO RP006-RUO
76634-908EA 549 USD InStock
CA76634-906 76634-908 CA76634-908 76634-904 76634-906 CA76634-904
Anti-HER2/neu (c-erb-2) Rabbit Polyclonal Antibody
Antibodies
This antibody recognizes a c-erbB-2 protein, which is a receptor tyrosine kinase of the c-erbB family. It is closely related in structure to the epidermal growth factor receptor. c-erbB-2 oncoprotein is detectable in a subset of breast and other adenocarcinomas, as well as transitional cell carcinomas. In the case of breast cancer, expression determined by immunohistochemistry has been shown to be associated with poor prognosis.
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty