Would you like to log in again?
Anti-Fibronectin Rabbit Polyclonal Antibody
Supplier: Diagnostic BioSystems
RP013-05-RUO RP013-01-RUO RP013-05 RP013-01-RUO RP013-RUO RP013
76634-938EA 1390.16 USD InStock
CA76634-936 76634-938 CA76634-938 76634-934 76634-936 CA76634-934
Anti-Fibronectin Rabbit Polyclonal Antibody
Antibodies
This antibody reacts with human fibronectin in liver, tonsil, skin and kidney. Traces of contaminating antibodies have been removed by solid-phase adsorption. Only the fibronectin arch was observed in crossed immunoelectrophoresis.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty