Would you like to log in again?
Anti-Helicobacter pylori Rabbit Polyclonal Antibody
Supplier: Diagnostic BioSystems
RP016-01-RUO RP016 RP016-05-RUO RP016-RUO RP016-01 RP016-05
76634-950EA 440.69 USD InStock
76634-950 CA76634-946 76634-948 CA76634-948 76634-946 CA76634-950
Anti-Helicobacter pylori Rabbit Polyclonal Antibody
Antibodies
This antibody reacts with the whole H. pylori. The antibody stains the bacteria present on the surface of the epithelium or in the cytoplasm of the epithelial cells. H. pylori is known to cause peptic ulcer and chronic gastritis in humans.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty