Would you like to log in again?
Anti-HSV I Rabbit Polyclonal Antibody
Supplier: Diagnostic BioSystems
RP018-05-RUO RP018-05 RP018-01 RP018-RUO RP018-01-RUO RP018
76634-956EA 494.44 USD InStock
CA76634-954 76634-956 CA76634-956 76634-952 76634-954 CA76634-952
Anti-HSV I Rabbit Polyclonal Antibody
Antibodies
This antibody reacts with HSV type I specific antigens and with antigens common to HSV type I and II virus. It reacts with all the major glycoproteins present in the viral envelope as well as with at least one core protein.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty