Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Anti-IgM Rabbit Polyclonal Antibody
Supplier: Diagnostic BioSystems
RP024-05 RP024-RUO RP024-05-RUO RP024-01-RUO RP024-RUO RP024-01
76634-982EA 121.82 USD InStock
76634-982 CA76634-986 76634-984 CA76634-982 CA76634-984 76634-986
Anti-IgM Rabbit Polyclonal Antibody
Antibodies
This antibody reacts with the heavy chain present in all types of human IgM. It stains B cell follicles in human lymphoid tissues. In lymphoid tissues, IgM containing plasma cells stain strongly but immune complexes are usually undetectable.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty