Would you like to log in again?
Anti-Lambda light chain Rabbit Polyclonal Antibody
Supplier: Diagnostic BioSystems
RP027-05-RUO RP027-01-RUO RP027-RUO RP027-05 RP027 RP027-01-RUO
76635-002EA 199.45 USD InStock
76635-002 CA76635-000 76635-000 CA76635-004 CA76635-002 76635-004
Anti-Lambda light chain Rabbit Polyclonal Antibody
Antibodies
This antibody does not show any cross-reactivity with kappa light chain. It stains B cell follicles in human lymphoid tissues. The mantle zone shows a mosaic pattern of labeling while the germinal centers show a coarse meshwork pattern of staining.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty