Would you like to log in again?
Anti-Gastrin Rabbit Polyclonal Antibody
Supplier: Diagnostic BioSystems
RP044-01-RUO RP044-RUO RP044-01 RP044-05 RP044 RP044-05-RUO
76635-032EA 297.38 USD InStock
CA76635-030 76635-032 76635-030 CA76635-034 76635-034 CA76635-032
Anti-Gastrin Rabbit Polyclonal Antibody
Antibodies
The antibody is specific against gastrin-containing cells. This anti-body reacts with human gastrin I and human gastrin I fragments. No cross-reactivity was observed with rat gastrin, human big gastrin, human gastrin releasing peptide, sulfated cholecystokinin amide, non-sulfated cholecystokinin amide and cholecystokinin.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty