Would you like to log in again?
Anti-Albumin Rabbit Polyclonal Antibody
Supplier: Diagnostic BioSystems
RP046-05 RP046-RUO RP046 RP046-01-RUO RP046-01 RP046-05-RUO
76635-040EA 231.69 USD InStock
CA76635-040 76635-040 76635-036 CA76635-038 76635-038 CA76635-036
Anti-Albumin Rabbit Polyclonal Antibody
Antibodies
This antibody reacts with human albumin. This antibody is essentially free of other rabbit serum proteins. Specificity is determined by Ouchterlony double diffusion (ODD) and immunoelectrophoresis (IEP) versus human serum and human albumin.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty