Would you like to log in again?
Anti-Alpha-1-antitrypsin Rabbit Polyclonal Antibody
Supplier: Diagnostic BioSystems
RP048-RUO RP048-01-RUO RP048 RP048-05-RUO RP048-01 RP048-05
76635-052EA 259.16 USD InStock
CA76635-052 76635-052 CA76635-050 CA76635-048 76635-048 76635-050
Anti-Alpha-1-antitrypsin Rabbit Polyclonal Antibody
Antibodies
The antibody is specific against human α-1-antitrypsin. This antibody stains macrophages in human tissues and does not stain any other cell type.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty