Would you like to log in again?
Anti-Myeloperoxidase Rabbit Polyclonal Antibody
Supplier: Diagnostic BioSystems
RP053-05 RP053-05-RUO RP053-01 RP053 RP053-RUO RP053-01-RUO
76635-068EA 332.01 USD InStock
76635-068 CA76635-066 76635-066 CA76635-068 CA76635-070 76635-070
Anti-Myeloperoxidase Rabbit Polyclonal Antibody
Antibodies
Myeloperoxidase metabolizes most of the hydrogen peroxide generated by activated phagocytes. This antibody stains strongly with the cytoplasm of myeloid cells. The antibody detects myeloperoxidase found in myeloid cells of acute myeloid leukemia. The myeloperoxidase gene has been localized to chromosome 17.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty