Would you like to log in again?
Anti-Mmp-9 Rabbit Polyclonal Antibody
Supplier: Diagnostic BioSystems
RP066 RP066-05-RUO RP066-05 RP066-01-RUO RP066-01 RP066-RUO
76635-086EA 546.99 USD InStock
76635-086 CA76635-084 76635-084 CA76635-088 76635-088 CA76635-086
Anti-Mmp-9 Rabbit Polyclonal Antibody
Antibodies
This antibody reacts with a 92 kDa pro form and 86 kDa active form of MMP-9. MMPs are proteolytic enzymes capable of degrading connective tissue components. MMPs play a crucial role in tumor cell invasion and metastasis.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty