Would you like to log in again?
Anti-Beta-Catenin Rabbit Polyclonal Antibody
Supplier: Diagnostic BioSystems
RP080-01 RP080-RUO RP080-05-RUO RP080-01-RUO RP080 RP080-05
76635-102EA 142.12 USD InStock
76635-102 CA76635-104 76635-106 CA76635-102 76635-104 CA76635-106
Anti-Beta-Catenin Rabbit Polyclonal Antibody
Antibodies
The catenins are structurally related cytoplasmic proteins which have been classified as alpha (α), beta (β), and gamma (γ), these proteins can bind to the highly conserved, intracellular cytoplasmic tail of E-cadherin. Together, the catenin/cadherin complexes play an important role mediating cellular adhesion. β-catenin associates with the cytoplasmic portion of E-cadherin, which is necessary for the function of E-cadherin as an adhesion molecule.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty